missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ GC Capillary Column Connectors: Replacement Ferrules
Replacement SilTite ferrules for SilTiteμ-Union
Brand: Trajan™ 073566
This item is not returnable.
View return policy
Description
- SilTite μ-Union gives low thermal mass - 9mm in length and <0.5g mass
- Zero dead volume for optimized peak shape
- Chemically deactivated stainless steel ferrules give system inertness
- Robust and stable for both high temperature and pressure cycling
Specifications
Replacement μ-Union Ferrule | |
Stainless Steel | |
0.1 to 0.25 mm I.D. First and Second Columns with 0.4 mm Holes |