Learn More
Trajan™ GC Inlet Liner Accessories
Used as sealing rings for Agilent GC.
Brand:  Trajan™ 0726532
Product Code. 11942637
  • 53.20 € / Pack of 10
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Suitable for Agilent GC
  • Temperature stable up to 300°C
Specifications
Specifications
Viton O-Ring
Viton
Viton O-ring for Agilent GC's
Agilent, Thermo trace 1300 Bruker (Varian) Except SG092235
10/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Trajan™ GC Inlet Liner Accessories > O-Ring, Viton; For All Liners Except SG092235