Learn More
SGE™ NanoVolume Syringes
Ideal for nanovolume capillary chromatography injection as well as preparing accurate standards which require small volumes.
Brand:  SGE™ 000310
 View more versions of this product
Product Code. 10482001
  • 133.00 € / Each
Explore more special offers
Description
Description
Ideal for nanovolume capillary chromatography injection as well as preparing accurate standards which require small volumes.
  • Ability to inject down to 50nL with high precision and accuracy
  • Ideal for dispensing very small liquid volumes
  • Plunger extends into the needle tip
  • Sample is contained only in the needle without any glass contact
  • Virtually zero dead volume and carry over between injections
  • Recommended for use when sample volume is less than 1000nL (1μL)
Specifications
Specifications
±2%
23 G
0.315 in.
0.63 mm
2.75 in.
NanoVolume Syringe
Ea.
Removable Cone
Borosilicate Glass (Barrel)
0.0248 in.
8 mm
2.75 in.
70 mm
0.5BR-7
0.5 μL
Removable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SGE™ NanoVolume Syringes > Tip Style: cone; Capacity: 0.5μL; Gauge: 23; O.D.: 0.63mm; Length: 70mm