Learn More
SGE™ Series II Standard Fixed Needle Syringes
Standard syringes with anti-roll flange and needles that are aligned with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
Brand:  SGE™ 001000
 View more versions of this product
Product Code. 10518502
  • 57.85 € / Each
Explore more special offers
Description
Description
Standard syringes with anti-roll flange and needles that are aligned with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
  • Stainless-steel plungers are hand fitted, noninterchangeable, nonreplaceable
  • Syringes with 5 and 10μL capacities have barrel O.D. of 6.5mm, scale length of 54.1mm and include a 50mm long 26s gauge needle
  • 25 to 500μL capacity syringes have 8mm O.D. barrel, 60mm scale length and 50mm long, 25 gauge needle Fixed-needle models have needle glued into barrel with solvent-resistant epoxy glue Removable-needle models feature a PTFE seal, a spring and nut for leak-resistant operation
Specifications
Specifications
±1%
26 G
0.25 in.
0.47 mm
1.96 in.
Major - 0.5 μL, Minor - 0.1 μL
Ea.
Fixed Bevel
Borosilicate Glass (Barrel)
0.01 in.
6.5 mm
Plunger protection
Microvolume Syringe
5F
5 μL
Fixed
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SGE™ Series II Standard Fixed Needle Syringes > Fixed needle; Needle: 50mm; Gauge: 26; O.D.: 0.47mm; Volume: 5μL; Ea.