missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs, aqua
Aid the process of speedy and reliable biopsy specimen labeling with Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs.
197.00 € - 234.00 €
Specifications
Pore Style | Small Square Pore |
---|---|
Writing Face Angle | 45° |
Product Type | Biopsy Cassette |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
12943765
|
Epredia™
B1000730AQ |
Aqua | N/A | N/A | N/A | |||||
12593926
|
Epredia™
B1000730BL |
Blue | N/A | N/A | N/A | |||||
12953765
|
Epredia™
B1000730FGR |
Fluorescent Green | N/A | N/A | N/A | |||||
12963765
|
Epredia™
B1000730FPK |
Fluorescent Pink | N/A | N/A | N/A | |||||
12973765
|
Epredia™
B1000730FYW |
Fluorescent Yellow | N/A | N/A | N/A | |||||
12583926
|
Epredia™
B1000730GR |
Green | N/A | N/A | N/A | |||||
12983765
|
Epredia™
B1000730GY |
Gray | N/A | N/A | N/A | |||||
12993765
|
Epredia™
B1000730LI |
Lilac | N/A | N/A | N/A | |||||
12903775
|
Epredia™
B1000730OR |
Orange | N/A | N/A | N/A | |||||
12913775
|
Epredia™
B1000730PE |
Peach | N/A | N/A | N/A | |||||
12573926
|
Epredia™
B1000730PK |
Pink | N/A | N/A | N/A | |||||
12923775
|
Epredia™
B1000730TN |
Tan | N/A | N/A | N/A | |||||
12563926
|
Epredia™
B1000730WH |
White | N/A | N/A | N/A | |||||
15135944
|
Epredia™
B1000730YW |
Yellow | N/A | N/A | N/A | |||||
Description
- Writing face is flat 45° angle
- Locking lid attached by two hinges
- Front or back opening
Ordering Information:
- Packaged in 75-cassette tubes
Specifications
Small Square Pore | |
45° | |
Biopsy Cassette |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs, aqua