Learn More
Epredia™ Cryochrome™ Embedding Resin
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
Brand:  Epredia™ 9990426
87.55 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 12570017
  • 106.00 € / Pack of 4
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Color codes frozen sections for easy identification and tracking
  • Four colors available: pink, blue, yellow, green
  • Formulated with physical characteristics identical to Cryomatrix™ embedding resin
Specifications
Specifications
Embedding Media
120mL