Learn More
Epredia™ Cytoblock™ Cell Block Preparation System
Facilitate quick, reliable processing of FNAs, tissue fragments, and cell buttons with the Cytoblock™ Cell Block Preparation System.
Brand:  Epredia™ 7401151
50.40 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 10446640
  • 61.50 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
  • Suitable for immunohistochemical techniques
  • Replaces time-consuming and costly agar and thrombin techniques
  • Requires phosphate-free fixative on processor, such as Epredia™ fixatives Zinc Formal-Fixx™, Formal-Fixx, or Glyo-Fixx™ (formalin substitute) or simply bypass formalin fixation steps on processor
  • Cytoblock system can also be run on a rapid biopsy program
Specifications
Specifications
Replacement Reagent
Cell Block Preparation System Reagent