Learn More
Epredia™ Cytoblock™ Cell Block Preparation System
Facilitate quick, reliable processing of FNAs, tissue fragments, and cell buttons with the Cytoblock™ Cell Block Preparation System.
60.00 € - 126.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10066588
|
Epredia™
7401150 |
Cell Preparation System |
126.00 €
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10446640
|
Epredia™
7401151 |
Replacement Reagent |
60.00 €
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
Description
- Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
- Suitable for immunohistochemical techniques
- Replaces time-consuming and costly agar and thrombin techniques
- Requires phosphate-free fixative on processor, such as Epredia™ fixatives Zinc Formal-Fixx™, Formal-Fixx, or Glyo-Fixx™ (formalin substitute) or simply bypass formalin fixation steps on processor
- Cytoblock system can also be run on a rapid biopsy program