missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Cytospin™ Collection Fluid
Store specimens or unstained slides for weeks without adverse effect with Epredia™ Shandon™ Cytospin™ Collection Fluid.
Brand: Epredia™ 6768315
52.92 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Packaged in convenient pump bottle or pre-filled specimen cups
- Ready-to-use alcohol/Carbowax™ (PEG) combination for direct specimen collection and fixation
Specifications
Collection Fluid | |
500mL | |
Alcohol/Carbowax (PEG) combination |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
- H370-Causes damage to organs.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P235-Keep cool.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : FD&C yellow No. 5, ethyl alcohol, isopropyl alcohol, methylalcohol, polyethylene glycol, methylene