missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Cytospin™ Collection Fluid
Store specimens or unstained slides for weeks without adverse effect with Epredia™ Shandon™ Cytospin™ Collection Fluid.
64.60 € - 385.00 €
Specifications
Product Type | Collection Fluid |
---|---|
Solution Type | Alcohol/Carbowax (PEG) combination |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12570057
|
Epredia™
9990323 |
120mL | N/A | ||||||
12638016
|
Epredia™
9990323 |
120mL |
385.00 €
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10178278
|
Epredia™
6768315 |
500mL |
64.60 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10589248
|
Epredia™
6768001 |
4 L |
77.90 €
3.80 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Packaged in convenient pump bottle or pre-filled specimen cups
- Ready-to-use alcohol/Carbowax™ (PEG) combination for direct specimen collection and fixation
Specifications
Collection Fluid | |
Alcohol/Carbowax (PEG) combination |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title