missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Cytospin™ Collection Fluid

Store specimens or unstained slides for weeks without adverse effect with Epredia™ Shandon™ Cytospin™ Collection Fluid.

Brand:  Epredia™ 6768001

63.84 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.



 View more versions of this product

Product Code. 10589248

  • 77.90 € / 3.80 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Packaged in convenient pump bottle or pre-filled specimen cups
  • Ready-to-use alcohol/Carbowax™ (PEG) combination for direct specimen collection and fixation
Specifications

Specifications

Collection Fluid
4 L
Alcohol/Carbowax (PEG) combination
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A
  • Flammable liquid Category 3
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H350-May cause cancer.
  • H370-Causes damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P235-Keep cool.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : FD&C yellow No. 5, ethyl alcohol, isopropyl alcohol, methylalcohol, polyethylene glycol, methylene

SDS
Documents

Documents

Certificates
Special Offers

Special Offers