Learn More
Epredia™ Double Cytoslide™ Microscope Slides
Use Epredia™ Shandon™ Double Cytoslides™ with Epredia™ Shandon™ EZ Double Cytofunnel™ Chambers.
Brand: Epredia™ 5991054
68.80 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Specimen is displayed within a circle marked as 1 or 2
- Number circles correspond with the Cytofunnel™ chamber
Ordering Alert:
- Sold in boxes of 100 slides
Specifications
Double Cytoslide | |
Plain | |
Noncoated |