Learn More
Epredia™ Shandon™ Gill™ Hematoxylin
Achieve fast and convenient results, while maintaining complete control over stain intensity with Epredia™ Shandon™ Gill™ Hematoxylin, which has mucoid staining.
Brand:  Epredia™ 6765010
 View more versions of this product
Product Code. 10188158
  • 156.00 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Gill 1: Light blue nuclei
  • Gill 2: Medium blue nuclei
  • Gill 3: Blue-black nuclei
Specifications
Specifications
Shandon Gill™ 3 Hematoxylin
4L