Learn More
Epredia™ Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads
Absorb and lock in liquids with Epredia™ Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads.
378.00 € - 1500.00 €
Product Code | Brand | Product Type | Length (English) | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Length (English) | Width (English) | Price | Quantity & Availability | |||
12552736
|
Epredia™
5641 |
Formaldehyde Neutralizer Pad | 10 in. | 8 in. |
1455.00 €
Pack of 400 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12532736
|
Epredia™
56411 |
Formaldehyde Neutralizer Pad | 10 in. | 8 in. |
378.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12542736
|
Epredia™
5643 |
Formaldehyde Neutralizer Pad | 20 in. | 16 in. | N/A | ||||
Description
- Fiber resists tearing, even when saturated
- Formaldehyde neutralizer pads reduce exposure by chemically bonding with formaldehyde, resulting in the encapsulation of vapors—ideal for grossing areas and autopsy work
- Clinical pads work as surgical cutting board pads, dissecting pads, autopsy table pads and liners for body trays and cadaver bags
Specifications
Formaldehyde |