missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Tissue-Marking Dyes, Tissue marking dye kit

Permanently mark the margins of excised surgical specimens for exact orientation with Epredia™ Shandon™ Tissue-Marking Dyes.

Brand:  Epredia™ 3120128

37.00 EUR valid until 2025-12-31
Use promo code "24374" to get your promotional price.



 View more versions of this product

Product Code. 10415339

  • 38.95 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Special binding pigment evenly coats the tissue without penetrating tissue surface
  • Does not interfere with diagnostic interpretation
  • Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning

Ordering Information:

Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).

Specifications

Specifications

Green
Tissue Marking Dye
59 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 59 ML GREEN TISSUE MARKING DYE, PERMANENT, MARK .

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P391-Collect spillage.

Supplemental information
  • MIXTURE LIST-Contains : Isopropanol, 2-(Dimethylamino) ethanol,Ammonium hydroxide,Formaldehyde

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.