Learn More
Epredia™ Shandon™ Tissue-Marking Dyes, Tissue marking dye kit
Permanently mark the margins of excised surgical specimens for exact orientation with Epredia™ Shandon™ Tissue-Marking Dyes.
Brand: Epredia™ 3120126
15.52 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
Description
- Special binding pigment evenly coats the tissue without penetrating tissue surface
- Does not interfere with diagnostic interpretation
- Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning
Ordering Information:
Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).
Specifications
Yellow | |
Tissue Marking Dye | |
59 mL | |
Healthcare |
Safety and Handling
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H371-May cause damage to organs.
Precautionary Statement
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P362-Take off contaminated clothing.
Supplemental information
- MIXTURE LIST-Contains : ammnoium hydroxide,formaldehyde, diarylide yellow AAOT
Your input is important to us. Please complete this form to provide feedback related to the content on this product.