missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Tissue-Marking Dyes, Tissue marking dye kit
Permanently mark the margins of excised surgical specimens for exact orientation with Epredia™ Shandon™ Tissue-Marking Dyes.
Brand: Epredia™ 3120125
This item is not returnable.
View return policy
Description
- Special binding pigment evenly coats the tissue without penetrating tissue surface
- Does not interfere with diagnostic interpretation
- Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning
Ordering Information:
Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).
Specifications
Black | |
Tissue Marking Dye | |
59 mL |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H350-May cause cancer.
- H370-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : carbon black, ammonium hydroxide, formaldehyde
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction