missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Xylene Substitute

Achieve safer, more convenient processing, staining and coverslipping with Epredia™ Xylene Substitute.

Brand:  Epredia™ 9990505

100.50 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.



 View more versions of this product

Product Code. 10128638

  • 161.00 € / 4 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Not sensitizing to normal skin
  • Evaporation rate similar to xylene
  • Virtually odorless aliphatic hydrocarbon—no citrus aroma
  • Airborne exposure limit three times safer than xylene (TLV 300ppm)
Specifications

Specifications

Xylene Substitute
1 gal.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Shandon Xylene Substitute

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H340-May cause genetic defects.
  • H350-May cause cancer.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers