Learn More
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
Brand:  Thermo Scientific™ RSK23SD5
 View more versions of this product
Product Code. 11349351
  • 1145.70 € / Each
Estimated Shipment: 05-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features
  • Stainless steel racks
  • Boxes
  • Cell dividers
Warranty varies by country. Please contact us for details.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100
Shelf Kit
Stainless Steel
HERAfreeze HFU B Series, Revco ExF and DxF Series
Shelf Kit
Boxes (80/5 Racks)
5
5
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers > Shelf Kit, Includes: 5 sliding drawer racks and (80) 2 in. boxes with 100 cell dividers, for use with HERAfreeze HFU B Series, Revco ExF and DxF Series