missing translation for 'onlineSavingsMsg'
Learn More
SHOWA NSK26 Nitrile Chemical Resistant Gloves Product Code.: 17871211
Add to basket

SHOWA NSK26 Nitrile Chemical Resistant Gloves

Product Code. 17871211
Size: Large
Click to view available options
Size: Large
This item is not returnable. View return policy

Product Code. 17871211

Brand: SHOWA NSK2610L

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

The SHOWA NSK26 glove is a dependable and effective means of safeguarding against a range of solvents, oils, and acids. It provides excellent protection against these potentially harmful substances.

  • Ensures excellent hand protection in hazardous environments.
  • The glove has a flexible and watertight construction.
  • It is designed to offer optimal protection to the wearer's hands.
Featuring a rough surface finish, the NSK26 offers excellent grip, allowing you to handle tools and materials with ease. The cotton liner wicks away sweat, ensuring maximum comfort even during extended use. Furthermore, an eyelet and elasticated cuff keep the glove in place, ensuring full and consistent protection against a range of hazards. Whether you work in the chemical, automotive, or manufacturing industry, the SHOWA NSK26 glove is an excellent choice for providing reliable chemical protection, grip, and comfort for your hands.

Specifications

Chlorinated No
Applications Industrial, Laboratory
Color Blue
Latex Free Yes
Material Nitrile
Size Large
EN388 Standard EN388:4111X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Lining Unlined
Length (English) 25.6 in.
Length (Metric) 650 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SHOWA NSK26 Nitrile Chemical Resistant Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.