missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Signature Series™ Bluing Reagent
Ensure crisp nuclear detail with the ready-to-use Epredia™ Signature Series Bluing Reagent.
Brand: Epredia™ 7301
This item is not returnable.
View return policy
Description
- Buffered to ensure proper alkalinity (pH = 8.0)
- Specially formulated to prevent tissue separation from the slide
- Requires only a one-minute water rinse versus conventional bluing reagents which require up-to five minutes
Available in 5 liter packaging
- Reduces plastic by 90%
- Increases stain stability
- Reduces messy stain spills
- Saves 35% storage space versus traditional packaging
Specifications
Bluing Reagent | |
1 gal. | |
8 | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction