missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Signature Series™ Bluing Reagent

Ensure crisp nuclear detail with the ready-to-use Epredia™ Signature Series Bluing Reagent.

18.50 € - 56.40 €

Specifications

Product Type Bluing Reagent
pH 8
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10862090
View Documents
Epredia™
7301
1 gal. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10259988
View Documents
Epredia™
7341
4 x 1 gal.
56.40 €
Pack of 4
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
Description

Description

  • Buffered to ensure proper alkalinity (pH = 8.0)
  • Specially formulated to prevent tissue separation from the slide
  • Requires only a one-minute water rinse versus conventional bluing reagents which require up-to five minutes

Available in 5 liter packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging
Specifications

Specifications

Bluing Reagent
8
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.