Learn More
Epredia™ Signature Series™ Clear-Rite 3™
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series Clear-Rite 3.
181.00 €
Specifications
Product Type Clearing Agent
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10138198
View Documents
Epredia™
6901
4 x 1 gal.
181.00 €
15.14 L
Description
Description
  • Very low odor; Benzene-free
  • Use in all tissue processors and automatic stainers
  • Does not produce hard or brittle specimens, even after prolonged exposure
Specifications
Specifications
Clearing Agent
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 1B
  • Acute toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H371-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : proprietary mixtureof aliphatic hydrocarbons