Learn More
BRAND™ SILBERBRAND Automatic Burette with PTFE Stockcock
SILBERBRAND Automatic Burette equipped with an i-stopcock and lateral stopcock.
Brand:  BRAND™ 22336
333.11 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10663012
  • 395.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Error limits according to class B, DIN EN ISO 385
  • Interchangeable keys
  • Calibrated to deliver (TD, Ex)
  • With glass-key in the titration and intermediate stopcock
  • Automatic zeroing
  • Intermediate stopcock to recirculate residual liquid
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mL
Without bottle
12 months
0.1 mL
i-stopcock, amber lateral stopcock, PTFE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products