missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ SILBERBRAND Compact Automatic Burette with Schellbach Stripe

Compact Automatic Burette designed to be fast to dismantle and easy to clean.

Brand:  BRAND™ 23811

115.80 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10330445

  • 134.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Easy to repair
  • All individual components are replaceable
  • Detachable PTFE stopcock
  • Calibrated to deliver (TD, Ex)
  • Error limit according to class B, DIN EN ISO 385
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.1 mL
Borosilicate Glass (3.3)
Black Schellbach Stripe, 1000 mL LDPE Bottle
12 months
Class B
50 mL
0.1 mL
PTFE
Automatic Buret
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.