Learn More
BRAND™ Silberbrand™ Eterna™ Class B Borosilicate Glass Mixing Cylinders
Manufactured from borosilicate glass and complies with DIN EN ISO 4788 standards. BRAND™ Silberbrand™ Eterna™ Class B Borosilicate Glass Mixing Cylinders include a polypropylene or polyethylene stopper and hexagonal base.
Brand:  BRAND™ 33920
41.99 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 10417523
  • 46.15 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from borosilicate glass
  • Complies with DIN EN ISO 4788 standards.
  • Calibrated to contain (TC, In).
  • Includes a polypropylene or polyethylene stopper and hexagonal base.
  • Height measured without stopper.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hexagonal
Amber
Graduated Mixing Cylinder
DIN EN ISO 4788
25 mL
Batch Certificate, Polypropylene Stopper
NS 14/23
0.5 mL
To Contain
Round
Borosilicate Glass
B
190 mm
Amber Graduations
Polypropylene
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Silberbrand™ Eterna™ Class B Borosilicate Glass Mixing Cylinders > Capacity (Metric): 25 mL