missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Silica gel, for column chromatography, Ultra Pur
For column chromatography.
146.00 € - 2588.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10407982
|
Thermo Scientific Chemicals
360050010 |
1 kg |
146.00 €
1kg |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
10619004
|
Thermo Scientific Chemicals
360050050 |
5 kg |
566.00 €
5kg |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
10500764
|
Thermo Scientific Chemicals
360050300 |
30 kg |
2588.00 €
30kg |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7631-86-9 |
Specifications
40 to 60 μm | |
Plastic Bottle | |
Silica Gel |
60 Å | |
Silica Gel | |
7631-86-9 |
RUO – Research Use Only