missing translation for 'onlineSavingsMsg'
Learn More

Silicate solution for Ion Chromatography 1g/L, 1% NaOH matrix, Fisher Chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

54.00 € - 114.00 €

Specifications

Concentration 1g/L
Solution Contains H2O

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12902195
View Documents
Fisher Chemical
A 041 100
100 mL
54.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
12912195
View Documents
Fisher Chemical
A 041 250
250 mL
77.00 €
250mL
Please to purchase this item. Need a web account? Register with us today!
12922195
View Documents
Fisher Chemical
A 041 500
500 mL
114.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

1g/L
H2O
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Single-component Standard Solution for Ion chromatography. Silicate 1000mg/l in NaOH 1% (A 041)

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

SDS
Documents

Documents

Product Certifications