Learn More
Silicon carbide, beta-phase, nanopowder, Thermo Scientific Chemicals
S.A. 70 to 90 m2/g
72.36 € - 700.49 €
Chemical Identifiers
CAS | 409-21-2 |
---|---|
Molecular Formula | CSi |
Molecular Weight (g/mol) | 40.10 |
MDL Number | MFCD00049531 |
InChI Key | HBMJWWWQQXIZIP-UHFFFAOYSA-N |
Synonym | silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide |
PubChem CID | 9863 |
ChEBI | CHEBI:29390 |
SMILES | [C-]#[Si+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435480
|
Thermo Scientific Alfa Aesar
044646.06 |
5 g |
72.36 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445480
|
Thermo Scientific Alfa Aesar
044646.14 |
25 g |
235.17 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455480
|
Thermo Scientific Alfa Aesar
044646.22 |
100 g |
700.49 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silicon carbide can be used form very hard ceramics that are widely used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide as light-emitting diodes (LEDs) and detectors in early radios.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
409-21-2 | |
40.10 | |
HBMJWWWQQXIZIP-UHFFFAOYSA-N | |
9863 | |
[C-]#[Si+] |
CSi | |
MFCD00049531 | |
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide | |
CHEBI:29390 |
Specifications
2700°C | |
Black to Gray | |
CSi | |
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide | |
HBMJWWWQQXIZIP-UHFFFAOYSA-N | |
methanidylidynesilylium | |
9863 | |
40.1 | |
Silicon carbide, beta-phase |
409-21-2 | |
Powder | |
MFCD00049531 | |
Insoluble in water | |
[C-]#[Si+] | |
40.10 | |
CHEBI:29390 | |
Odorless |
Safety and Handling
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H350
missing translation for 'einecsNumber' : 206-991-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.