Learn More
Saint Gobain Life Sciences Silicon Folding Skirt Stopper
Use this stopper for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Silicon Folding Skirt Stopper is particularly suited for all types of tissue culture work.
Brand:  Saint Gobain Life Sciences 467018-20
 View more versions of this product
Product Code. 11714608
  • 37.10 € / Pack of 20
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Two-part folding skirt stoppers
  • Designed with a double seal to keep container contents secure
  • Ideal for securing glass, metal and plastic containers
  • The lower section grips the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim
  • Upper hollow plug section can be perforated to extract contents without removing the stopper and remains watertight even after several perforations
  • Biologically neutral and non-toxic silicone caps for closing tubes and flasks
  • Odorless, tasteless, non-adherent and resistant to aging
  • Nontoxic, biologically neutral
  • FDA/BGA approved
  • For test tubes
  • Easy to install
TRUSTED_SUSTAINABILITY
Specifications
Specifications
16 mm
Transparent
Silicone Stopper
1 Ea.
Silicone
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Saint Gobain Life Sciences Silicon Folding Skirt Stopper > Silicon Folding Skirt Stopper; Diameter: 16 mm