Learn More
Lonza™ PYROGENT™ Gel Clot LAL Assay Kit
Performs qualitative LAL test for gram-negative bacterial endotoxin. Lonza PYROGENT™ Gel Clot LAL Assay Kit comes with 0.125EU/mL sensitivity.
Brand:  Lonza™ N194-125
Product Code. 13261994
  • 1885.00 € / Each
Explore more special offers
Description
Description
  • Provides simple positive/negative result for endotoxin testing
  • Gel clot assay is run in tubes and placed in water bath or dry heat block at 37°C
  • After one hour incubation period, tubes are flipped 180 degree
  • Firm clot in bottom of tube indicates positive reaction
  • If liquid flows down side of tube indicates negative for endotoxin
  • Kit do not include matched control standard endotoxin
  • Lysate kit with 0.125EU/mL sensitivity
  • Certificates of analysis is provided
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 Tests
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lonza™ PYROGENT™ Gel Clot LAL Assay Kit > 250 tests