missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, 40% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals

Identificatori chimici

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantity Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantity Prezzo Quantità e disponibilità  
11397517
missing translation for 'viewDocuments' Dettagli della promozione
Thermo Scientific Alfa Aesar
012727.A1
1 kg N/A
N/A
11307527
missing translation for 'viewDocuments' Dettagli della promozione
Thermo Scientific Alfa Aesar
012727.A7
5 kg N/A
N/A
Descrizione

Descrizione

Functionalized RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene; acrylate and acrylamide monomers. Azide group can be used to conjugate to a variety of alkyne-functionalized biomolecules. Chain Transfer Agent (CTA). SDS mixture of sodium alkyl sulfates consisting chiefly of sodium lauryl sulfate. Used as a suspending and thickening agent in pharmaceutical

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifica

Identificatori chimici

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
O=[Si]=O
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563

Specifica

7631-86-9
Particles in Liquid
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Insoluble in water.
O=[Si]=O
60.08
CHEBI:30563
Odorless
Silicon(IV) oxide, 40% in H2O
Colorless
O2Si
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
24261
60.09
1.3 g/cm3 at 20°C
Videos
Sicurezza e movimentazione

Sicurezza e movimentazione

Take precautionary measures against static discharges.

Flammable.

missing translation for 'einecsNumber' : 231-545-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documenti

Documenti

Certificati

RUO – Research Use Only