missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, Amorphous fumed, Surface treated, S.A. 105 to 130m2/g, Thermo Scientific Chemicals

For Use in Coatings, Adhesives, Sealants

34.97 € - 140.70 €

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11395607
View Documents
Thermo Scientific Alfa Aesar
042784.18
50 g
34.97 €
50g
Please to purchase this item. Need a web account? Register with us today!
11385607
View Documents
Thermo Scientific Alfa Aesar
042784.30
250 g
91.46 €
250g
Please to purchase this item. Need a web account? Register with us today!
11375607
View Documents
Thermo Scientific Alfa Aesar
042784.A1
1 kg
140.70 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used as sodium stabilizing counterion, for preparation of silica catalytic supports. Also used in glass manufacturing, foundry sand casting, metallurgical flux sand, ceramics sand, gas recovery, paint and coatings, filtration sand, chromatographic column treatment, wood treatment, inks, desiccants and powder metal molding. Its is principle gateway in electronic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 105 to 130m2/g
Specifications

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
O=[Si]=O
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563

Specifications

7631-86-9
Powder
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Insoluble in water.
O=[Si]=O
60.08
CHEBI:30563
Odorless
Silicon(IV) oxide
White
O2Si
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.09
1.8 g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

Wear suitable protective clothing.

H373: May cause damage to organs through prolonged or repeated exposure.

missing translation for 'einecsNumber' : 231-545-4

missing translation for 'rtecsNumber' : EU8655000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.