Learn More
Silicon powder, crystalline, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
55.58 € - 466.32 €
Chemical Identifiers
CAS | 7440-21-3 |
---|---|
Molecular Formula | Si |
Molecular Weight (g/mol) | 28.09 |
MDL Number | MFCD00085311 |
InChI Key | XUIMIQQOPSSXEZ-UHFFFAOYSA-N |
Synonym | metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 |
PubChem CID | 5461123 |
ChEBI | CHEBI:27573 |
IUPAC Name | silicon |
SMILES | [Si] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397507
|
Thermo Scientific Alfa Aesar
012681.09 |
10 g |
55.58 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387507
|
Thermo Scientific Alfa Aesar
012681.22 |
100 g |
159.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377507
|
Thermo Scientific Alfa Aesar
012681.36 |
500 g |
466.32 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-21-3 | |
28.09 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
5461123 | |
silicon |
Si | |
MFCD00085311 | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
CHEBI:27573 | |
[Si] |
Specifications
7440-21-3 | |
99.5% (Metals basis) | |
MFCD00085311 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
silicon | |
5461123 | |
Odorless | |
Silicon |
Crystalline Powder | |
Si | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
[Si] | |
28.09 | |
CHEBI:27573 | |
Powder |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-130-8
missing translation for 'rtecsNumber' : VW0400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.