Learn More
Heidolph instruments Silicone Covers
Flexible and chemically resistant protection against splashes and dripping water for MR Hei-Mix D, MR Hei-Mix L, MR Hei-Tec und MR Hei-End.
Brand:  Heidolph instruments 23-07-06-05-59
Product Code. 10497163
  • 21.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Replaceable cover against splashes and dripping water for MR Hei-Tec,MR Hei-Connect and MR Hei-End
  • Silicone protective cover
  • Flexible and chemically resistant
  • Protection against splashes and dripping water for MR Hei-Mix D, MR Hei-Mix L, MR Hei-Tec and MR Hei-End
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For HEI-MIX/TEC/END
Silicone
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Heidolph instruments Silicone Covers > For HEI-MIX/TEC/END