Learn More
Fisherbrand™ Silicone Gasket
Accessory for Mini-Plus Horizontal Gel Electrophoresis Unit
Brand:  Fisherbrand™ HU-SG
Product Code. 10132131
Description
Description
  • Silicone gaskets
  • Lines casting tray
  • Forms leak proof seal against inner walls of running chamber when the casting tray is turned at 90°
Specifications
Specifications
Fisherbrand Mini-Plus Horizontal Gel Electrophoresis Unit
Gasket
1 Each
1 m
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Silicone Gasket > Silicone