missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Silicone Sealing Ring, VMQ

The silicone seal is used for sealing and fixing threaded pipes with threaded caps.

Brand:  DWK Life Sciences 292350607

46.27 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11820720

  • 52.40 € / Pack of 10
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The silicone sealing ring matches the screw connection piece and has good sealing properties. Due to the PTFE layer on one side of the sealing ring, there is a certain inertness of the material. The included hole allows the insertion of glass rods, olives, hoses, etc.

  • With bonded PTFE face
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5.5 to 6.5 mm
GL 18
Fits Tubing ID: 5.5 to 6.5 mm
Silicone (VMQ)
Sealing; Connecting glass tubes; Suitable for PBT screw cap with aperture
10/Pk.
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers