Learn More
Macherey-Nagel™ Silicone Septa
Brand:  Macherey-Nagel™ 702606
 View more versions of this product
Product Code. 10358341
  • 65.95 € / Pack of 50
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Septum
45° Shore A
Clear
50 Pack
13 mm
250°C
Silicone