Learn More
Fisherbrand™ Silicone Stoppers
For laboratory and/or food testing, industrial masking or educational use.
10.55 € - 35.00 €
Product Code | Brand | Diameter (Metric) Bottom | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Bottom | Quantity | Price | Quantity & Availability | |||||
11527662
|
Fisherbrand™
SB013/S |
13 mm | 10/Pk. |
19.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10131511
|
Fisherbrand™
SB013/S |
13 mm | 10/Pk. | N/A | ||||||
11547662
|
Fisherbrand™
SB017/S |
17 mm | 10/Pk. |
25.00 €
Pack of 10 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
10040131
|
Fisherbrand™
SB017/S |
17 mm | 10/Pk. | N/A | ||||||
11517672
|
Fisherbrand™
SB031/S |
31 mm | 2/Pk. |
20.00 €
Pack of 2 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10060141
|
Fisherbrand™
SB031/S |
31 mm | 2/Pk. | N/A | ||||||
10283601
|
Fisherbrand™
SB003/S |
3 mm | 50/Pk. | N/A | ||||||
11537652
|
Fisherbrand™
SB003/S |
3 mm | 50/Pk. |
14.00 €
Pack of 50 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
11577672
|
Fisherbrand™
SB043/S |
43 mm | 1/Pk. |
23.00 €
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
10181841
|
Fisherbrand™
SB043/S |
43 mm | 1/Pk. | N/A | ||||||
11597672
|
Fisherbrand™
SB049/S |
49 mm | 1/Pk. |
29.00 €
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
10599561
|
Fisherbrand™
SB049/S |
49 mm | 1/Pk. | N/A | ||||||
11507682
|
Fisherbrand™
SB053/S |
53 mm | 1/Pk. |
35.00 €
Each |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
10440693
|
Fisherbrand™
SB053/S |
53 mm | 1/Pk. | N/A | ||||||
11567652
|
Fisherbrand™
SB006/S |
6 mm | 50/Pk. |
17.00 €
Pack of 50 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
10757311
|
Fisherbrand™
SB006/S |
6 mm | 50/Pk. | N/A | ||||||
11587652
|
Fisherbrand™
SB008/S |
8 mm | 20/Pk. |
11.00 €
Pack of 20 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
10213751
|
Fisherbrand™
SB008/S |
8 mm | 20/Pk. | N/A | ||||||
Description
- Non-toxic, translucent
- Temperature range -60°C to 200°C
- High electrical resistance
- High resistance to many chemicals including ozone
Specifications
Silicone | |
Stopper |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Silicone Stoppers