missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Silicone Stoppers
For laboratory and/or food testing, industrial masking or educational use.
Brand: Fisherbrand™ SB017/S
Description
- Non-toxic, translucent
- Temperature range -60°C to 200°C
- High electrical resistance
- Resists cracking and spallation
- High resistance to many chemicals including ozone
Specifications
10/Pk. | |
17 mm | |
Silicone | |
Stopper |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Silicone Stoppers > Diameter: 17mm
Spot an opportunity for improvement?Share a Content Correction