Learn More
Buerkle™ Silo Drill
If bulk goods are transported over long distances (in trucks, railway goods wagons), separation may occur.Taking the sample just from the surface therefore leads to the wrong result.Representative samples from all layers of the silo can be taken with the SiloDrill.
Brand:  Buerkle™ 5301-2000
3936.51 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10607055
  • 4340.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Representative samples from all layers of a storage or materials silo can be taken with the SiloDrill. The SiloDrill fits through most standard openings with a diameter of 90mm.
  • Standard length 150 cm, extendable in increments of 100 cm, max. 350 cm
  • Sprung inner tube prevents jamming of coarse grains
  • Scraper edge for cohesive substances
  • Sturdy, removable T-handle
  • Drill tip with drilling blades for easy boring
  • Easy removal of the sample chamber, simple cleaning
Specifications
Specifications
Stainless Steel (AISI 304)
400 mL
150 cm
Autoclavable
40 mm