Learn More
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Use SilTite GC Capillary Column Connectors for fused silica capillary GC columns.
Brand: Thermo Scientific™ 290MU499
112.19 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Use Thermo Scientific™ SilTite Capillary Column Connectors for fused silica capillary GC columns. They are glass lined for inertness and have a low thermal mass.
Join together two GC columnsRepair broken columns
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capillary Column Connector | |
290 MU499 | |
32 mm | |
Fused Silica Capillary Column |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.