missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SilTite™ GC Capillary Column Connector

Use SilTite GC Capillary Column Connectors for fused silica capillary GC columns.

86.37 € - 112.75 €

Specifications

For Use With (Equipment) Fused Silica Capillary Column
View More Specs

Products 4
Product Code Brand Type For Use With (Equipment) Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Type For Use With (Equipment) Diameter (Metric) Inner Price Quantity & Availability  
10497514
View Documents
Thermo Scientific™
290MU498
Capillary Column Connector Fused Silica Capillary Column 0.1 to 0.25 mm
109.30 €
Each
In Stock
to see stock.
Add to basket
 
10117864
View Documents
Thermo Scientific™
290MU499
Capillary Column Connector Fused Silica Capillary Column 32 mm
112.75 €
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
10301055
View Documents
Thermo Scientific™
290MU500
Capillary Column Connector Fused Silica Capillary Column 0.45 to 0.53 mm
112.75 €
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
10046604
View Documents
Thermo Scientific™
290MN211
Replacement Column Connector Nuts Fused Silica Capillary Column 0.1 to 0.53 mm
86.37 €
Pack of 5
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
Description

Description

Use Thermo Scientific™ SilTite Capillary Column Connectors for fused silica capillary GC columns. They are glass lined for inertness and have a low thermal mass.

  • Join two GC columns
  • Repair broken columns
Includes
One connector, two nuts and five ferrules.

Ordering Information
Five-pack of nuts also available. 
Specifications

Specifications

Fused Silica Capillary Column
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.