missing translation for 'onlineSavingsMsg'
Learn More
Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted, Thermo Scientific Chemicals Product Code.: 15485085

Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted, Thermo Scientific Chemicals

Product Code. 15485085
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 15485085

Brand: Thermo Scientific Alfa Aesar 047280.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as pharmaceutical intermediate and in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
Synonym Silver chloride dispersion in water
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Color White
Density 1.038 g/mL
UN Number UN3082
Quantity 500 g
Merck Index 14,8509
Solubility Information Dispersible in water. Very slightly soluble in cold water,soluble in solutions of alkali cyanides,thiosulfates,ammonium carbonates. Appreciably soluble in concentrated aqueous solutions of ammonium chloride,mercuric nitrate,and silver nitrate.
Formula Weight 143.32
Concentration or Composition (by Analyte or Components) 4.9% in H2O
Odor Odorless
Particle Size <200nm APS Number Weighted
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Silver chloride, Nanoparticle dispersion
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW23A040 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number047280.22,047280.36
Lot NumberS16H036 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number047280.22,047280.36
Lot NumberW23A039 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number047280.22,047280.36
Lot NumberP29H066 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number047280.22,047280.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.