missing translation for 'onlineSavingsMsg'
Learn More

Silver Conductive Ink, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 045661.06

32.70 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



 View more versions of this product

Product Code. 15492427

  • 32.86 € / 5g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Silver Conductive Ink
MFCD01868501
Liquid
2.5
5 g
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ12K010 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number045661.14,045661.06,045661.22
Lot NumberT01K046 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number045661.14,045661.06,045661.22
Lot NumberW12I003 Certificate TypeCertificate of Analysis Date16/09/2022 Catalog Number045661.14,045661.06,045661.22
Lot NumberT06I077 Certificate TypeCertificate of Analysis Date19/07/2022 Catalog Number045661.14,045661.06,045661.22
Lot NumberZ20C076 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number045661.14,045661.06,045661.22
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver Conductive Ink, Thermo Scientific Chemicals > 5g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only