missing translation for 'onlineSavingsMsg'
Learn More
Silver powder, spherical, APS 0.6-2 micron, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11368728

Silver powder, spherical, APS 0.6-2 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11368728
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11368728

Brand: Thermo Scientific Alfa Aesar 041598.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: S.A. 0.4-1.5m2/g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored powder
Density 10.5g/cm3
Assay Percent Range 99.9% (Metals basis)
Quantity 50 g
Odor Characteristic
Form Spherical Powder
Chemical Name or Material Silver

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM06L036 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog Number041598.09,041598.30,041598.18
Lot NumberQ25K007 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number041598.09,041598.30,041598.18
Lot NumberY19G040 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number041598.09,041598.30,041598.18,041600.18
Lot NumberS28I004 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog Number041598.09,041598.30,041598.18,041600.18
Lot NumberY30H028 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number041597.09,041597.30,041597.18,041598.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver powder, spherical, APS 0.6-2 micron, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.