missing translation for 'onlineSavingsMsg'
Learn More
Silver rod, 6.35mm (0.25in) dia x 150mm (5.9in) long, hard, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 15424755

Silver rod, 6.35mm (0.25in) dia x 150mm (5.9in) long, hard, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15424755
1 Pc.
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy

Product Code. 15424755

Brand: Thermo Scientific Alfa Aesar 013304.KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-22-4
Molecular Formula Ag
MDL Number MFCD00003397
Quantity 1 Pc.
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
SMILES [Ag]
IUPAC Name silver
Molecular Weight (g/mol) 107.87
PubChem CID 23954
ChEBI CHEBI:9141
Percent Purity 99.99%
Grade Premion™
Odor Characteristic
Weight ≈3.32g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Silver rod, 6.35mm (0.25 in.) dia. x 150mm (5.9 in.) long, Hard
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU11I043 Certificate TypeCertificate of Analysis Date17/08/2022 Catalog Number013304.KG
Lot NumberD14N31 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013304.KG
Lot NumberM19D017 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013304.KG
Lot NumberZ03E074 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013304.KG
Lot NumberP21D002 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number013304.KG

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver rod, 6.35mm (0.25in) dia x 150mm (5.9in) long, hard, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.