missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Silylating Silyl-991
Brand: Macherey-Nagel™ 701490.201
This item is not returnable.
View return policy
Specifications
Silylating Silyl-991 | |
1 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- Silyl-991, 20x1 mL
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Supplemental information
- MIXTURE LIST-Contains : N ,O-bis-trimethylsilyl-trifluoroacetamide (BSTFA), trimethylchlorosilane (TMCS), chlorotrimethylsil
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Silylating Silyl-991 > Quantity: 1mL
Spot an opportunity for improvement?Share a Content Correction