Learn More
Vitlab™ Simplex II Bottletop Dispenser
Vitlab™ Simplex II Bottletop Dispenser is designed to be practically universally applicable and can be used with many organic and inorganic solutions.
Brand:  Vitlab™ 1621508
 View more versions of this product
Product Code. 15600925
  • 697.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Direct displacement plunger with broad PFA seal prevents crystallisation build-up
  • Settings are quickly and accurately adjusted due to interior scalloped track and practical screw mechanism
  • Screwable ventilation plug for easy connection of a drying tube
  • Screwable discharge valve is equipped with an additional safety bulb and closes the dosing channel, if dispensing tube is not installed, so that no medium can escape
  • Equipped with a positive displacement piston and a fluoroplastic (PFA) sealing lip on the cylinder wall
  • Nominal volume 2 to 10mL includes adapters GL 25, GL 28, GL 32, GL 38, S 40 and telescopic intake tube (length 125 to 240mm)
  • Nominal volume 25 to 100mL includes adapters GL 32, GL 38, S 40 and telescopic intake tube (length 170 to 330mm)
Specifications
Specifications
10.0 to 100.0 mL