Learn More
Vitlab™ Simplex II Bottletop Dispenser
Vitlab™ Simplex II Bottletop Dispenser is designed to be practically universally applicable and can be used with many organic and inorganic solutions.
361.00 € - 697.00 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15670915
|
Vitlab™
1621503 |
0.2 to 2.0 mL |
361.00 €
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
15624854
|
Vitlab™
1621504 |
0.5 to 5.0 mL |
361.00 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
15634854
|
Vitlab™
1621505 |
1.0 to 10.0 mL |
361.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
15680915
|
Vitlab™
1621506 |
2.5 to 25 mL |
472.00 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
15600925
|
Vitlab™
1621508 |
10.0 to 100.0 mL |
697.00 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
Description
- Direct displacement plunger with broad PFA seal prevents crystallisation build-up
- Settings are quickly and accurately adjusted due to interior scalloped track and practical screw mechanism
- Screwable ventilation plug for easy connection of a drying tube
- Screwable discharge valve is equipped with an additional safety bulb and closes the dosing channel, if dispensing tube is not installed, so that no medium can escape
- Equipped with a positive displacement piston and a fluoroplastic (PFA) sealing lip on the cylinder wall
- Nominal volume 2 to 10mL includes adapters GL 25, GL 28, GL 32, GL 38, S 40 and telescopic intake tube (length 125 to 240mm)
- Nominal volume 25 to 100mL includes adapters GL 32, GL 38, S 40 and telescopic intake tube (length 170 to 330mm)